CAS RN 1034352-30-1
IUPAC Name:
2-bromo-3-(2-ethylhexyl)-5-iodothiophene
Synonyms:
2-bromo-3-(2-ethylhexyl)-5-iodothiophene
Molecular Weight (g/mol):
401.14
Molecular Formula:
C12H18BrIS
InChi Key:
VHAVRRSIFMAOTI-UHFFFAOYNA-N
SMILES:
CCCCC(CC)CC1=C(Br)SC(I)=C1
1
–
1
of
1
results
1
2-Bromo-3-(2-ethylhexyl)-5-iodothiophene (stabilized with Copper chip) 97.0+%, TCI America™
CAS: 1034352-30-1 Molecular Formula: C12H18BrIS Molecular Weight (g/mol): 401.144 InChI Key: VHAVRRSIFMAOTI-UHFFFAOYSA-N PubChem CID: 70976734 IUPAC Name: 2-bromo-3-(2-ethylhexyl)-5-iodothiophene SMILES: CCCCC(CC)CC1=C(SC(=C1)I)Br