accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1034449-15-4

IUPAC Name: 1,3-dicyclohexyl-3H-1λ⁵,3-benzodiazol-1-ylium chloride
Synonyms: 1,3-dicyclohexyl-1λ⁵,3-benzodiazol-1-ylium chloride
Molecular Weight (g/mol): 318.89
Molecular Formula: C19H27ClN2
InChi Key: JACUDTKAZHCIPU-UHFFFAOYSA-M
SMILES: [Cl-].C1CCC(CC1)N1C=[N+](C2CCCCC2)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)