accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNN

CAS RN 1034566-07-8

IUPAC Name: methyl[(1-phenyl-1H-imidazol-2-yl)methyl]amine
Synonyms: methyl[(1-phenylimidazol-2-yl)methyl]amine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: IZYNHWOVGJNYQF-UHFFFAOYSA-N
SMILES: CNCC1=NC=CN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results