accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOCl

CAS RN 1034566-09-0

IUPAC Name: 3,4-dihydro-2H-1-benzopyran-8-carbonyl chloride
Synonyms: 3,4-dihydro-2H-1-benzopyran-8-carbonyl chloride
Molecular Weight (g/mol): 196.63
Molecular Formula: C10H9ClO2
InChi Key: TTYLUAYKWAEQDM-UHFFFAOYSA-N
SMILES: ClC(=O)C1=C2OCCCC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results