accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3H3CNSiCH3CH3H3CH3CSiCH3H3CNSiCH3CH3H3CH3CSiCH3H3CNSiCH3CH3H3CEr

CAS RN 103457-72-3

IUPAC Name: erbium(3+) tris(bis(trimethylsilyl)azanide)
Synonyms: erbium(3+) tris(bis(trimethylsilyl)azanide)
Molecular Weight (g/mol): 648.42
Molecular Formula: C18H54ErN3Si6
InChi Key: ZVNGWTZZRANJAO-UHFFFAOYSA-N
SMILES: [Er+3].C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results