accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103470-68-4

IUPAC Name: yttrium(3+) tris(2-ethylhexanoate)
Synonyms: yttrium(3+) tris(2-ethylhexanoate)
Molecular Weight (g/mol): 518.52
Molecular Formula: C24H45O6Y
InChi Key: AGOMHFKGCMKLDA-UHFFFAOYNA-K
SMILES: [Y+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)