CAS RN 103478-58-6
IUPAC Name:
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-methylbutanoic acid
Synonyms:
(2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-3-methylbutanoic acid|FMOC-N-METHYL-D-VALINE
Molecular Weight (g/mol):
353.42
Molecular Formula:
C21H23NO4
InChi Key:
YCXXXPZNQXXRIG-LJQANCHMSA-N
SMILES:
CC(C)[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O