accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHCH3CH3

CAS RN 10348-47-7

IUPAC Name: ethyl 2-hydroxy-4-methylpentanoate
Synonyms: ethyl 2-hydroxy-4-methylpentanoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: QRHOWVDPHIXNEN-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)CC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results