accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNOOHOOH

CAS RN 10351-75-4

IUPAC Name: 1H-1,3-benzodiazole-5,6-dicarboxylic acid
Synonyms: 1H-1,3-benzodiazole-5,6-dicarboxylic acid
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H6N2O4
InChi Key: PIPQOFRJDBZPFR-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C2N=CNC2=C1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)