accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1035391-48-0

IUPAC Name: tert-butyl N-{[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbamate
Synonyms: tert-butyl N-{[2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbamate
Molecular Weight (g/mol): 351.23
Molecular Formula: C18H27BFNO4
InChi Key: VAYYEUXFQWTMJJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=C(F)C=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results