accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3NBOOH3CH3CH3CH3C

CAS RN 1035631-57-2

IUPAC Name: 9-phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 495.23
Molecular Formula: C30H35B2NO4
InChi Key: BLKLIQJTGNDTOL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(C=C(C=C1)B1OC(C)(C)C(C)(C)O1)N2C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results