accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClOHOHN

CAS RN 10357-84-3

IUPAC Name: hydrogen 6-hydroxy-1,2-dihydropyridin-2-one chloride
Synonyms: hydrogen 6-hydroxy-2(1H)-pyridinone chloride
Molecular Weight (g/mol): 147.56
Molecular Formula: C5H6ClNO2
InChi Key: HNWWAWKDVFVJRG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC1=CC=CC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results