CAS RN 10361-12-3
CAS RN 10361-12-3
IUPAC Name:
octadecyl 3,4,5-trihydroxybenzoate
Synonyms:
octadecyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol):
422.61
Molecular Formula:
C25H42O5
InChi Key:
BRNPAEUKZMBRLQ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1
1
–
1
of
1
results
1
Stearyl Gallate 97.0+%, TCI America™

CAS: 10361-12-3 Molecular Formula: C25H42O5 Molecular Weight (g/mol): 422.61 MDL Number: MFCD00016429 InChI Key: BRNPAEUKZMBRLQ-UHFFFAOYSA-N Synonym: Gallic Acid Stearyl Ester PubChem CID: 473591 IUPAC Name: octadecyl 3,4,5-trihydroxybenzoate SMILES: CCCCCCCCCCCCCCCCCCOC(=O)C1=CC(O)=C(O)C(O)=C1