CAS RN 10361-37-2
Barium Chloride Anhydrous (Laboratory), Fisher Chemical™
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, 10% w/v aq. soln.
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, 0.5N Standardized Solution
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, 0.1N Standardized Solution
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, ultra dry, 99.998% (metals basis)
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, anhydrous, 99.998% (metals basis)
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 SMILES: [Cl-].[Cl-].[Ba++]
Barium Chloride, Anhydrous, Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium AA Standard, 1000 ppm Ba in 3% HCl, Ricca Chemical
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
VeriSpec™ Barium (Ba) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
VeriSpec™ Barium Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium AA Standard, 1000 ppm Ba in 3% HNO3, Ricca Chemical
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium AA Standard, Certified, 1000ppm Ba ±10ppm (1mL = 1mg Ba), LabChem™
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+) dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride solution, 20%, Solstice
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
- 1
- 2