accessibility menu, dialog, popup

UserName

Viewing profile as user:
BiONOOONOOONOO

CAS RN 10361-44-1

IUPAC Name: bismuth(3+) trinitrate
Synonyms: bismuth(III) nitrate
Molecular Weight (g/mol): 394.99
Molecular Formula: BiN3O9
InChi Key: PPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES: [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results