accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOBiO

CAS RN 10361-46-3

IUPAC Name: (nitrooxy)bismuthanone
Synonyms: bismuth oxynitrate
Molecular Weight (g/mol): 286.98
Molecular Formula: BiNO4
InChi Key: OGCYGKRWXWQOAK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)O[Bi]=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results