accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10361-84-9

IUPAC Name: scandium(3+) trichloride
Synonyms: scandium chloride
Molecular Weight (g/mol): 151.31
Molecular Formula: Cl3Sc
InChi Key: DVMZCYSFPFUKKE-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Sc+3]
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)