accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103661-13-8

IUPAC Name: 4-(4-hydroxyphenyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(4-hydroxyphenyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 227.28
Molecular Formula: C10H13NO3S
InChi Key: NSHOLIWPDBJMPT-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N1CCS(=O)(=O)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results