accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSNH2

CAS RN 103694-26-4

IUPAC Name: 1-(2-methyl-1,3-thiazol-4-yl)methanamine
Synonyms: 1-(2-methyl-1,3-thiazol-4-yl)methanamine
Molecular Weight (g/mol): 128.19
Molecular Formula: C5H8N2S
InChi Key: ZCKAEFOHSOQKHN-UHFFFAOYSA-N
SMILES: CC1=NC(CN)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results