accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10375-34-5

IUPAC Name: furan-2,5-dicarbonyl dichloride
Synonyms: furan-2,5-dicarbonyl dichloride
Molecular Weight (g/mol): 192.98
Molecular Formula: C6H2Cl2O3
InChi Key: PDSULNVJASBMLP-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(O1)C(Cl)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results