accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10377-58-9

IUPAC Name: magnesium(2+) diiodide
Synonyms: magnesium iodide
Molecular Weight (g/mol): 278.11
Molecular Formula: I2Mg
InChi Key: BLQJIBCZHWBKSL-UHFFFAOYSA-L
SMILES: [Mg++].[I-].[I-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)