accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHOOHONOHOOOHH2ONaNaNa

CAS RN 10378-22-0

IUPAC Name: 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid hydrate trisodium
Synonyms: edetic acid hydrate trisodium
Molecular Weight (g/mol): 379.23
Molecular Formula: C10H18N2Na3O9
InChi Key: DFCJTQJARZUURB-UHFFFAOYSA-N
SMILES: O.[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results