accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10378-49-1

IUPAC Name: sodium tetrabromogolduide
Synonyms: sodium tetrabromoaurate|sodium tetrabromoaurate(III)|Na[AuBr4]|sodium gold bromide|sodium tetrabromoaurate(1-)
Molecular Weight (g/mol): 539.57
Molecular Formula: AuBr4Na
InChi Key: PMPPMRNISYJZFW-UHFFFAOYSA-J
SMILES: [Na+].Br[Au-](Br)(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results