accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10378-50-4

IUPAC Name: 4-[4,7-bis(benzyloxy)-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl]-N,N-bis(propan-2-yl)butanamide
Synonyms: 4-[4,7-bis(benzyloxy)-1,3-dioxoisoindol-2-yl]-N,N-diisopropylbutanamide
Molecular Weight (g/mol): 528.65
Molecular Formula: C32H36N2O5
InChi Key: PXMOBROHNZPLTM-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)CCCN1C(=O)C2=C(OCC3=CC=CC=C3)C=CC(OCC3=CC=CC=C3)=C2C1=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)