accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103788-65-4

IUPAC Name: 2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol
Synonyms: 2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol
Molecular Weight (g/mol): 203.24
Molecular Formula: C12H13NO2
InChi Key: JYWHQBLLIBQGCU-UHFFFAOYSA-N
SMILES: CC1=C(CCO)N=C(O1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results