accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103836-71-1

IUPAC Name: 1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-one
Synonyms: 1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-one
Molecular Weight (g/mol): 370.29
Molecular Formula: C19H12F6O
InChi Key: OIUBVYQZMUKRRF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=CC(=O)C=CC2=CC=C(C=C2)C(F)(F)F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results