accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 10387-13-0

IUPAC Name: 9,10-bis(chloromethyl)anthracene
Synonyms: 9,10-bis(chloromethyl)anthracene
Molecular Weight (g/mol): 275.17
Molecular Formula: C16H12Cl2
InChi Key: UOSROERWQJTVNU-UHFFFAOYSA-N
SMILES: ClCC1=C2C=CC=CC2=C(CCl)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results