accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10387-40-3

IUPAC Name: ethanethioic S-acid potassium
Synonyms: thioacetic acid, potassium salt
Molecular Weight (g/mol): 115.21
Molecular Formula: C2H4KOS
InChi Key: SDJHDRMYZQFJJO-UHFFFAOYSA-N
SMILES: [K].CC(S)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (4)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)