accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10389-51-2

IUPAC Name: 4-(4-nitrophenyl)morpholine
Synonyms: morpholine, 4-(4-nitrophenyl)-
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: IAJDSUYFELYZCS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results