accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3NHOOCH3CH3H3COOCH3CH3(E)

CAS RN 103890-78-4

IUPAC Name: 3,5-diethyl 4-{2-[(1E)-3-(tert-butoxy)-3-oxoprop-1-en-1-yl]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: lacidipine glaxosmithkline brand of lacidipine lacidipine (product) lacidipinum lacidipino motens boehringer ingelheim brand of lacidipine menarini brand of lacidipine 拉西地平 lacipil
Molecular Weight (g/mol): 455.55
Molecular Formula: C26H33NO6
InChi Key: GKQPCPXONLDCMU-CCEZHUSRSA-N
SMILES: CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1\C=C\C(=O)OC(C)(C)C)C(=O)OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results