accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNH2

CAS RN 10394-38-4

IUPAC Name: 1-methyl-1H-1,3-benzodiazol-5-amine
Synonyms: 1-methyl-1,3-benzodiazol-5-amine
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: IWBGBYZGEQUDBT-UHFFFAOYSA-N
SMILES: CN1C=NC2=CC(N)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results