accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFI

CAS RN 103962-05-6

IUPAC Name: 1-iodo-4-(trifluoromethoxy)benzene
Synonyms: 1-iodo-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 288.01
Molecular Formula: C7H4F3IO
InChi Key: RTUDBROGOZBBIC-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(I)C=C1
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)