accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3ONOHN(R)(R)SNOHOOSOOSH2NN(Z)

CAS RN 103980-44-5

IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride
Synonyms: ceftiofur hydrochloride
Molecular Weight (g/mol): 560.01
Molecular Formula: C19H18ClN5O7S3
InChi Key: KEQFDTJEEQKVLM-JUODUXDSSA-N
SMILES: Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results