accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHNOO

CAS RN 104-04-1

IUPAC Name: N-(4-nitrophenyl)acetamide
Synonyms: nitroacetanilide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: NQRLPDFELNCFHW-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)