accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-19-8

IUPAC Name: dimethyl[2-(4-methylpiperazin-1-yl)ethyl]amine
Synonyms: dimethyl[2-(4-methylpiperazin-1-yl)ethyl]amine
Molecular Weight (g/mol): 171.29
Molecular Formula: C9H21N3
InChi Key: XFLSMWXCZBIXLV-UHFFFAOYSA-N
SMILES: CN(C)CCN1CCN(C)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results