accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOCl

CAS RN 104-29-0

IUPAC Name: 3-(4-chlorophenoxy)propane-1,2-diol
Synonyms: chlorphenesin maolate gechophen mycil C9H11ClO3 chlorphenesine demykon chlorphenesin preparation 2-chlorphenesin chlorphenesin (substance)
Molecular Weight (g/mol): 202.63
Molecular Formula: C9H11ClO3
InChi Key: MXOAEAUPQDYUQM-UHFFFAOYNA-N
SMILES: OCC(O)COC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)