accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 104-36-9

IUPAC Name: 1,4-dibutoxybenzene
Synonyms: benzene, 1,4-dibutoxy-
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: ZROGCHDPRZRKTI-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(OCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results