accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-49-4

IUPAC Name: 1,4-diisocyanatobenzene
Synonyms: benzene, 1,4-diisocyanato-
Molecular Weight (g/mol): 160.13
Molecular Formula: C8H4N2O2
InChi Key: ALQLPWJFHRMHIU-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)