accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 104-51-8

IUPAC Name: butylbenzene
Synonyms: N-butylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: OCKPCBLVNKHBMX-UHFFFAOYSA-N
SMILES: CCCCC1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)