accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-54-1

IUPAC Name: (2Z)-3-phenylprop-2-en-1-ol
Synonyms: styrone cinnamyl alcohol, (E)-isomer 3-fenyl-2-propen-1-ol cinnamic alcohol (natural) titanium (4+) salt cinnamyl alcohol (e)-isomer cinnamyl alcohol 2-propen-y1-ol, 3-phenyl- (e)-cinnamyl alcohol cinnamyl alcohol (product) cinnamyl alcohol (substance)
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: OOCCDEMITAIZTP-DAXSKMNVSA-N
SMILES: OC\C=C/C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)