accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OO

CAS RN 104-61-0

IUPAC Name: (5S)-5-pentyloxolan-2-one
Synonyms: gamma-nonalactone fema number 2781 coconut aldehyde aldehyde C-18 1,4-nonalolide 4-nonanolide gamma-nonyllactone 4-pentyl-butanolide gamma-amyl-gamma-butyrolactone nonan-4-olide
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: OALYTRUKMRCXNH-QMMMGPOBSA-N
SMILES: CCCCC[C@H]1CCC(=O)O1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)