accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-72-3

IUPAC Name: decylbenzene
Synonyms: 1-phenyldecane benzene, C10-13-alkyl derivs.
Molecular Weight (g/mol): 218.38
Molecular Formula: C16H26
InChi Key: UZILCZKGXMQEQR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)