accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-79-0

IUPAC Name: diethyl[2-(methylamino)ethyl]amine
Synonyms: diethyl[2-(methylamino)ethyl]amine
Molecular Weight (g/mol): 130.24
Molecular Formula: C7H18N2
InChi Key: MKDYQLJYEBWUIG-UHFFFAOYSA-N
SMILES: CCN(CC)CCNC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results