accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-83-6

IUPAC Name: 1-chloro-4-(chloromethyl)benzene
Synonyms: 4-chlorobenzal chloride
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: JQZAEUFPPSRDOP-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
  • (4)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (4)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (2)
  • (4)
  • (3)
  • (2)
  • (6)
  • (5)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)