accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-88-1

IUPAC Name: 4-chlorobenzaldehyde
Synonyms: 4-chlorobenzaldehyde ccris 857 C7H5ClO
Molecular Weight (g/mol): 140.57
Molecular Formula: C7H5ClO
InChi Key: AVPYQKSLYISFPO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=O)C=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (6)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)