accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-91-6

IUPAC Name: 4-nitrosophenol
Synonyms: 4-nitrosophenol P-nitrosophenol
Molecular Weight (g/mol): 123.11
Molecular Formula: C6H5NO2
InChi Key: JSTCPNFNKICNNO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results