accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104-94-9

IUPAC Name: 4-methoxyaniline
Synonyms: P-anisidine p-anisidine antimonyl tartrate
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: BHAAPTBBJKJZER-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1
Boiling Point 
  • (3)
  • (4)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (5)
Percent Purity 
  • (2)
  • (10)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (6)
  • (3)
  • (4)
  • (3)
  • (2)
  • (7)
  • (5)
  • (2)
  • (10)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)