accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSBr

CAS RN 104-95-0

IUPAC Name: 1-bromo-4-(methylsulfanyl)benzene
Synonyms: 1-bromo-4-(methylsulfanyl)benzene
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: YEUYZNNBXLMFCW-UHFFFAOYSA-N
SMILES: CSC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (8)
  • (3)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)