accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHN(E)

CAS RN 104-98-3

IUPAC Name: (2E)-3-(1H-imidazol-5-yl)prop-2-enoic acid
Synonyms: urocanic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: LOIYMIARKYCTBW-OWOJBTEDSA-N
SMILES: OC(=O)\C=C\C1=CN=CN1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results