accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NIOHOIHOO

CAS RN 1041-01-6

IUPAC Name: 2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
Synonyms: 3,5-diiodothyronine 3,5-diiodo-dl-thryronine
Molecular Weight (g/mol): 525.08
Molecular Formula: C15H13I2NO4
InChi Key: ZHSOTLOTTDYIIK-UHFFFAOYNA-N
SMILES: NC(CC1=CC(I)=C(OC2=CC=C(O)C=C2)C(I)=C1)C(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)